Stampa…

2-(Bromomethyl)-4-(trifluoromethyl)benzothiophene

Fornitore: Apollo Scientific
Pericolo

PC57794-100MG PC57794-1G PC57794-250MG
APOSPC57794-100MGEA 32.1 EUR
APOSPC57794-100MG APOSPC57794-1G APOSPC57794-250MG
2-(Bromomethyl)-4-(trifluoromethyl)benzothiophene
2-(Bromomethyl)-4-(trifluoromethyl)benzothiophene
Formula: C₁₀H₆BrF₃S
Peso molecolare: 295,119 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD31567869
CAS Number: 2377846-69-8
UN: 3261
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR