Stampa…

2-Bromo-6-fluoronicotinaldehyde 95%

Fornitore: Apollo Scientific

PC57508-1G PC57508-250MG
APOSPC57508-1GEA 720 EUR
APOSPC57508-1G APOSPC57508-250MG
2-Bromo-6-fluoronicotinaldehyde 95%
2-Bromo-6-fluoronicotinaldehyde
Formula: C₆H₃BrFNO
Peso molecolare: 203,997 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD16606508
CAS Number: 1227489-86-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR