Stampa…

2-Bromo-3-(trifluoromethoxy)aniline 99%

Fornitore: Apollo Scientific
Pericolo

PC56769-1G PC56769-250MG
APOSPC56769-1GEA 928 EUR
APOSPC56769-1G APOSPC56769-250MG
2-Bromo-3-(trifluoromethoxy)aniline 99%
2-Bromo-3-(trifluoromethoxy)aniline
Formula: C₇H₅BrF₃NO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28048416
CAS Number: 1805474-26-3
UN: 2811
ADR: 6.1,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR