Stampa…

2,3-Diamino-5-fluorobenzoic acid 95%

Fornitore: Apollo Scientific

PC56244-1G PC56244-250MG
APOSPC56244-1GEA 720 EUR
APOSPC56244-1G APOSPC56244-250MG
2,3-Diamino-5-fluorobenzoic acid 95%
2,3-Diamino-5-fluorobenzoic acid
Formula: C₇H₇FN₂O₂
Peso molecolare: 170,141 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12197351
CAS Number: 177960-38-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR