Stampa…

Methyl 2-bromo-5-(difluoromethoxy)benzoate

Fornitore: Apollo Scientific
Attenzione

PC56115-1G PC56115-250MG
APOSPC56115-1GEA 973 EUR
APOSPC56115-1G APOSPC56115-250MG
Methyl 2-bromo-5-(difluoromethoxy)benzoate
Methyl 2-bromo-5-(difluoromethoxy)benzoate
Formula: C₉H₇BrF₂O₃
Peso molecolare: 281.051 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD27927650
CAS Number: 705279-03-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR