2',4'-Difluoro-4-hydroxybiphenyl 98%
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
4-(2,4-Difluorophenyl)phenol, 2',4'-Difluoro-[1,1'-biphenyl]-4-ol
PC5378-1G
PC5378-5G
APOSPC5378-1GEA
161
EUR
APOSPC5378-1G
APOSPC5378-5G
2',4'-Difluoro-4-hydroxybiphenyl 98%
2',4'-Difluoro-4-hydroxybiphenyl
Formula:
C₁₂H₈F₂O Peso molecolare: 206,188 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03414359 CAS Number: 59089-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...