Stampa…

Methyl 4-chloro-2-fluoro-5-nitro-benzoate 98%

Fornitore: Apollo Scientific

PC53609-1G PC53609-250MG PC53609-5G
APOSPC53609-1GEA 168 EUR
APOSPC53609-1G APOSPC53609-250MG APOSPC53609-5G
Methyl 4-chloro-2-fluoro-5-nitro-benzoate 98%
Methyl 4-chloro-2-fluoro-5-nitro-benzoate
Formula: C₈H₅ClFNO₄
Peso molecolare: 233,581 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD12172989
CAS Number: 1070893-15-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR