Stampa…

Methyl 4-amino-3-(trifluoromethoxy)benzoate 95%

Fornitore: Apollo Scientific

PC53602-25G PC53602-5G
APOSPC53602-25GEA 764 EUR
APOSPC53602-25G APOSPC53602-5G
Methyl 4-amino-3-(trifluoromethoxy)benzoate 95%
Methyl 4-amino-3-(trifluoromethoxy)benzoate
Formula: C₉H₈F₃NO₃
Peso molecolare: 235,16 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD15527567
CAS Number: 457097-93-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR