Stampa…

5-Bromo-2,4-difluorobenzeneboronic acid 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: (5-Bromo-2,4-difluorophenyl)boronic acid, 2,4-Difluoro-5-bromophenylboronic acid

PC53563-1G PC53563-5G PC53563-25G
APOSPC53563-1GEA 65.5 EUR
APOSPC53563-1G APOSPC53563-5G APOSPC53563-25G
5-Bromo-2,4-difluorobenzeneboronic acid 98%
5-Bromo-2,4-difluorobenzeneboronic acid
Formula: C₆H₄BBrF₂O₂
Peso molecolare: 236,807 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD18783127
CAS Number: 2096331-76-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR