Stampa…

Tetramethylammonium (trifluoromethyl)sulphanide

Fornitore: Apollo Scientific
Attenzione

Synonyms: Tetramethylammonium trifluoromethanethiolate, Tetramethylazanium trifluoromethanethiolate

PC53546-1G PC53546-250MG
APOSPC53546-1GEA 584 EUR
APOSPC53546-1G APOSPC53546-250MG
Tetramethylammonium (trifluoromethyl)sulphanide
Tetramethylammonium (trifluoromethyl)sulphanide
Formula: CF₃S·C₄H₁₂N
Peso molecolare: 175,216 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD24842708
CAS Number: 515823-30-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR