Stampa…

1-Fluoronaphthalene-2-carboxylic acid 95%

Fornitore: Apollo Scientific

Synonyms: 1-Fluoro-2-naphthoic acid

PC53519-100MG PC53519-1G PC53519-250MG
APOSPC53519-100MGEA 161 EUR
APOSPC53519-100MG APOSPC53519-1G APOSPC53519-250MG
1-Fluoronaphthalene-2-carboxylic acid 95%
1-Fluoronaphthalene-2-carboxylic acid
Formula: C₁₁H₇FO₂
Peso molecolare: 190,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16990867
CAS Number: 574-97-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR