Stampa…

2,4-Difluoro-5-iodobenzyl bromide 95%

Fornitore: Apollo Scientific
Pericolo

PC53420-1G PC53420-250MG PC53420-5G
APOSPC53420-1GEA 161 EUR
APOSPC53420-1G APOSPC53420-250MG APOSPC53420-5G
2,4-Difluoro-5-iodobenzyl bromide 95%
2,4-Difluoro-5-iodobenzyl bromide
Formula: C₇H₄BrF₂I
Peso molecolare: 332,912 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD30474660
CAS Number: 2022670-54-6
UN: 3265
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR