2-(Difluoromethoxy)-5-fluorobenzaldehyde
Fornitore: Apollo Scientific
|
Attenzione
|
PC53174-1G
PC53174-250MG
APOSPC53174-1GEA
466
EUR
APOSPC53174-1G
APOSPC53174-250MG
2-(Difluoromethoxy)-5-fluorobenzaldehyde
2-(Difluoromethoxy)-5-fluorobenzaldehyde
Formula:
C₈H₅F₃O₂ Peso molecolare: 190,119 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD14698502 CAS Number: 1214326-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...