Stampa…

4-(2-Fluorobenzyl)piperidine hydrochloride 97%

Fornitore: Apollo Scientific

PC530027-1G PC530027-500MG
APOSPC530027-1GEA 524 EUR
APOSPC530027-1G APOSPC530027-500MG
4-(2-Fluorobenzyl)piperidine hydrochloride 97%
4-(2-Fluorobenzyl)piperidine hydrochloride
Formula: C₁₂H₁₆FN·ClH
Peso molecolare: 229,721 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09787475
CAS Number: 193357-26-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR