Boc-(R)-3-Amino-3-(3-fluorophenyl)propionic acid 97%
Fornitore: Apollo Scientific
Formula:
C₁₄H₁₈FNO₄ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03427956 CAS Number: 500789-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...