5-Fluoro-2-(trifluoromethyl)-1H-indole 95%
Fornitore: Apollo Scientific
PC52226-1G
PC52226-250MG
PC52226-5G
APOSPC52226-1GEA
285
EUR
APOSPC52226-1G
APOSPC52226-250MG
APOSPC52226-5G
5-Fluoro-2-(trifluoromethyl)-1H-indole 95%
5-Fluoro-2-(trifluoromethyl)-1H-indole
Formula:
C₉H₅F₄N Peso molecolare: 203,136 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD19689276 CAS Number: 1007235-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...