Stampa…

3-Bromobenzenesulphonyl fluoride 95%

Fornitore: Apollo Scientific

Synonyms: 3-Bromobenzene-1-sulphonyl fluoride

PC520754-1G PC520754-5G
APOSPC520754-1GEA 164 EUR
APOSPC520754-1G APOSPC520754-5G
3-Bromobenzenesulphonyl fluoride 95%
3-Bromobenzenesulphonyl fluoride
Formula: C₆H₄BrFO₂S
Peso molecolare: 239,062 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD22053310
CAS Number: 454-65-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR