Stampa…

4-Fluoropiperidine-1-sulphonyl chloride 95%

Fornitore: Apollo Scientific

PC520745-250MG PC520745-500MG
APOSPC520745-250MGEA 861 EUR
APOSPC520745-250MG APOSPC520745-500MG
4-Fluoropiperidine-1-sulphonyl chloride 95%
4-Fluoropiperidine-1-sulphonyl chloride
Formula: C₅H₉ClFNO₂S
Peso molecolare: 201.647 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18917110
CAS Number: 1224929-82-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR