Stampa…

Butyl-[1-(2-chloro-6-fluorophenyl)methylidine]amine 97%

Fornitore: Apollo Scientific

PC520625-10G PC520625-50G
APOSPC520625-10GEA 170 EUR
APOSPC520625-10G APOSPC520625-50G
Butyl-[1-(2-chloro-6-fluorophenyl)methylidine]amine 97%
Butyl-[1-(2-chloro-6-fluorophenyl)methylidine]amine
Formula: C₁₁H₁₃ClFN
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10699103
CAS Number: 945408-07-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR