Stampa…

3-Bromo-5-chloro-2-fluorobenzonitrile 97%

Fornitore: Apollo Scientific

PC51881-1G PC51881-250MG PC51881-5G
APOSPC51881-1GEA 495 EUR
APOSPC51881-1G APOSPC51881-250MG APOSPC51881-5G
3-Bromo-5-chloro-2-fluorobenzonitrile 97%
3-Bromo-5-chloro-2-fluorobenzonitrile
Formula: C₇H₂BrClFN
Peso molecolare: 234,453 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11846061
CAS Number: 1160574-15-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR