Stampa…

Trifluoromethyl 4-methylbenzenesulphonate 95%

Fornitore: Apollo Scientific

PC510358-100MG PC510358-1G PC510358-250MG
APOSPC510358-100MGEA 80 EUR
APOSPC510358-100MG APOSPC510358-1G APOSPC510358-250MG
Trifluoromethyl 4-methylbenzenesulphonate 95%
Trifluoromethyl 4-methylbenzenesulphonate
Formula: C₈H₇F₃O₃S
Peso molecolare: 240,2 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD31695793
CAS Number: 175676-42-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR