Stampa…

5,6-Difluorobenzimidazole

Fornitore: Apollo Scientific

PC510221-1G PC510221-250MG
APOSPC510221-1GEA 111 EUR
APOSPC510221-1G APOSPC510221-250MG
5,6-Difluorobenzimidazole
5,6-Difluorobenzimidazole
Formula: C₇H₄F₂N₂
Peso molecolare: 154,117 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02031530
CAS Number: 78581-99-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR