Stampa…

3,6-Difluoropyrazine-2-carbonitrile

Fornitore: Apollo Scientific

PC50486-100MG PC50486-1G PC50486-250MG
APOSPC50486-100MGEA 168 EUR
APOSPC50486-100MG APOSPC50486-1G APOSPC50486-250MG
3,6-Difluoropyrazine-2-carbonitrile
3,6-Difluoropyrazine-2-carbonitrile
Formula: C₅HF₂N₃
Peso molecolare: 141,078 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD22199270
CAS Number: 356783-28-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR