Stampa…

Ethyl 6-fluorobenzofuran-2-carboxylate

Fornitore: Apollo Scientific
Attenzione

PC50381-1G PC50381-250MG
APOSPC50381-1GEA 274 EUR
APOSPC50381-1G APOSPC50381-250MG
Ethyl 6-fluorobenzofuran-2-carboxylate
Ethyl 6-fluorobenzofuran-2-carboxylate
Formula: C₁₁H₉FO₃
Peso molecolare: 208,186 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28956404
CAS Number: 1089681-84-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR