Stampa…

3-Bromo-5-fluorobenzyl fluoride 95%

Fornitore: Apollo Scientific
Pericolo

PC502900-1G PC502900-250MG
APOSPC502900-1GEA 561 EUR
APOSPC502900-1G APOSPC502900-250MG
3-Bromo-5-fluorobenzyl fluoride 95%
3-Bromo-5-fluorobenzyl fluoride
Formula: C₇H₅BrF₂
Peso molecolare: 207,015 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD30829524
CAS Number: 1715032-20-4
UN: 3265
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR