Stampa…

2-Bromo-3-iodobenzyl fluoride 95%

Fornitore: Apollo Scientific
Pericolo

PC502885-1G PC502885-250MG
APOSPC502885-1GEA 611 EUR
APOSPC502885-1G APOSPC502885-250MG
2-Bromo-3-iodobenzyl fluoride 95%
2-Bromo-3-iodobenzyl fluoride
Formula: C₇H₅BrFI
Peso molecolare: 314,921 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD31634830
CAS Number: 2244085-66-1
UN: 3265
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR