Stampa…

2-Bromo-3-methoxybenzyl fluoride 95%

Fornitore: Apollo Scientific
Pericolo

PC502882-1G PC502882-250MG
APOSPC502882-1GEA 611 EUR
APOSPC502882-1G APOSPC502882-250MG
2-Bromo-3-methoxybenzyl fluoride 95%
2-Bromo-3-methoxybenzyl fluoride
Formula: C₈H₈BrFO
Peso molecolare: 219,051 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28531722
CAS Number: 1780793-90-9
UN: 3265
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR