2-(Difluoromethoxy)-4-fluorophenol 95%
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₇H₅F₃O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18395022 CAS Number: 1261572-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...