Stampa…

2,6-Dichloro-4-fluorobenzyl alcohol

Fornitore: Apollo Scientific

PC502274-1G PC502274-500MG
APOSPC502274-1GEA 458 EUR
APOSPC502274-1G APOSPC502274-500MG
2,6-Dichloro-4-fluorobenzyl alcohol
2,6-Dichloro-4-fluorobenzyl alcohol
Formula: C₇H₅Cl₂FO
Peso molecolare: 195,018 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28133674
CAS Number: 1806353-99-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR