Stampa…

3-Bromo-5-(difluoromethyl)pyridin-2-amine 95+%

Fornitore: Apollo Scientific
Pericolo

PC502272-1G PC502272-250MG
APOSPC502272-1GEA 1090 EUR
APOSPC502272-1G APOSPC502272-250MG
3-Bromo-5-(difluoromethyl)pyridin-2-amine 95+%
3-Bromo-5-(difluoromethyl)pyridin-2-amine
Formula: C₆H₅BrF₂N₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD25476827
CAS Number: 1803571-23-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR