Stampa…

5-Bromo-2-(trifluoromethyl)pyridine-3-carbaldehyde 95%

Fornitore: Apollo Scientific

PC50188-1G PC50188-250MG
APOSPC50188-1GEA 0 EUR
APOSPC50188-1G APOSPC50188-250MG
5-Bromo-2-(trifluoromethyl)pyridine-3-carbaldehyde 95%
5-Bromo-2-(trifluoromethyl)pyridine-3-carbaldehyde
Formula: C₇H₃BrF₃NO
Peso molecolare: 254,004 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD15528708
CAS Number: 1211536-83-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR