Stampa…

4-Amino-3-bromo-5-fluorobenzotrifluoride 95%

Fornitore: Apollo Scientific
Pericolo

Synonyms: 2-Bromo-6-fluoro-4-(trifluoromethyl)aniline

PC501453-1G PC501453-250MG
APOSPC501453-1GEA 768 EUR
APOSPC501453-1G APOSPC501453-250MG
4-Amino-3-bromo-5-fluorobenzotrifluoride 95%
4-Amino-3-bromo-5-fluorobenzotrifluoride
Formula: C₇H₄BrF₄N
Peso molecolare: 258,011 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28397844
CAS Number: 1034325-63-7
UN: 2810
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR