3-Chloro-5-fluorobenzene -1,2-diamine
Fornitore: Apollo Scientific
Synonyms:
3-Chloro-5-fluorobenzene-1,2-diamine
PC501051-1G
PC501051-250MG
APOSPC501051-1GEA
195
EUR
APOSPC501051-1G
APOSPC501051-250MG
3-Chloro-5-fluorobenzene -1,2-diamine
3-Chloro-5-fluorobenzene -1,2-diamine
Formula:
C₆H₆ClFN₂ Peso molecolare: 160,577 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD20723094 CAS Number: 153505-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...