Stampa…

2-Fluororesorcinol

Fornitore: Apollo Scientific

Synonyms: 2-Fluorobenzene-1,3-diol

PC500671-1G PC500671-250MG
APOSPC500671-1GEA 239 EUR
APOSPC500671-1G APOSPC500671-250MG
2-Fluororesorcinol
2-Fluororesorcinol
Formula: C₆H₅FO₂
Peso molecolare: 128,101 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09835187
CAS Number: 103068-40-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR