Potassium (2,2-dimethylprop-1-yl)trifluoroborate 98%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
Potassium trifluoro(neopentyl)borate
PC50057-1G
PC50057-5G
APOSPC50057-1GEA
109
EUR
APOSPC50057-1G
APOSPC50057-5G
Potassium (2,2-dimethylprop-1-yl)trifluoroborate 98%
Potassium (2,2-dimethylprop-1-yl)trifluoroborate
Formula:
C₅H₁₁BF₃·K Peso molecolare: 178,045 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09271780 CAS Number: 1150655-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...