Stampa…

4-Bromo-5-fluoro-2-iodoaniline 95+%

Fornitore: Apollo Scientific

PC500251-1G PC500251-5G
APOSPC500251-1GEA 132 EUR
APOSPC500251-1G APOSPC500251-5G
4-Bromo-5-fluoro-2-iodoaniline 95+%
4-Bromo-5-fluoro-2-iodoaniline
Formula: C₆H₄BrFIN
Peso molecolare: 315,91 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD26386356
CAS Number: 1219741-79-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR