(2-Chloro-6-fluorophenyl)methanesulphonyl chloride
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
(2-Chloro-6-fluorophenyl)methanesulphonyl chloride, 2-Chloro-6-fluorobenzylsulphonyl chloride
PC50016-1G
PC50016-250MG
APOSPC50016-1GEA
121
EUR
APOSPC50016-1G
APOSPC50016-250MG
(2-Chloro-6-fluorophenyl)methanesulphonyl chloride
(2-Chloro-6-fluorophenyl)methanesulphonyl chloride
Formula:
C₇H₅Cl₂FO₂S Peso molecolare: 243,083 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09045976 CAS Number: 926257-07-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...