Stampa…

3-Bromo-5-methoxybenzotrifluoride

Fornitore: Apollo Scientific

Synonyms: 1-Bromo-3-methoxy-5-(trifluoromethyl)benzene, 1-Bromo-3-(trifluoromethyl)-5-methoxybenzene

PC499010-1G PC499010-25G PC499010-5G
APOSPC499010-1GEA 36.4 EUR
APOSPC499010-1G APOSPC499010-25G APOSPC499010-5G
3-Bromo-5-methoxybenzotrifluoride
3-Bromo-5-methoxybenzotrifluoride
Formula: C₈H₆BrF₃O
Peso molecolare: 255,032 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD09999639
CAS Number: 627527-23-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR