4-Fluoro-2-nitrothiophenol 95%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
4-Fluoro-2-nitrophenyl mercaptan, 4-Fluoro-2-nitrobenzenethiol
PC49621-1G
PC49621-250MG
APOSPC49621-1GEA
257
EUR
APOSPC49621-1G
APOSPC49621-250MG
4-Fluoro-2-nitrothiophenol 95%
4-Fluoro-2-nitrothiophenol
Formula:
C₆H₄FNO₂S Peso molecolare: 173,165 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD22481385 CAS Number: 654063-68-0 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...