Stampa…

1-Chloro-4-(1,1,1-trifluoroprop-2-yl)benzene 95%

Fornitore: Apollo Scientific

Synonyms: 2-(4-Chlorophenyl)-1,1,1-trifluoropropane

PC49560-10G PC49560-1G PC49560-5G
APOSPC49560-10GEA 1700 EUR
APOSPC49560-10G APOSPC49560-1G APOSPC49560-5G
1-Chloro-4-(1,1,1-trifluoroprop-2-yl)benzene 95%
1-Chloro-4-(1,1,1-trifluoroprop-2-yl)benzene
Formula: C₉H₈ClF₃
Peso molecolare: 208,608 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD14525518
CAS Number: 1204295-79-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR