4-Cyclopropyl-2-fluoroaniline
Fornitore: Apollo Scientific
|
Attenzione
|
PC49416-1G
PC49416-250MG
PC49416-5G
APOSPC49416-1GEA
430
EUR
APOSPC49416-1G
APOSPC49416-250MG
APOSPC49416-5G
4-Cyclopropyl-2-fluoroaniline
4-Cyclopropyl-2-fluoroaniline
Formula:
C₉H₁₀FN Peso molecolare: 151,181 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD06802502 CAS Number: 893739-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...