3',5'-Dichloro-2,2,2,4'-tetrafluoroacetophenone
Fornitore: Apollo Scientific
Formula:
C₈H₂Cl₂F₄O |
MDL Number:
MFCD23382507 CAS Number: 1190865-44-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...