Stampa…

(2,3-Difluorophenyl)methanesulphonyl chloride

Fornitore: Apollo Scientific
Pericolo

Synonyms: α-(Chlorosulphonyl)-2,3-difluorotoluene

PC49121-1G PC49121-500MG PC49121-250MG
APOSPC49121-1GEA 379 EUR
APOSPC49121-1G APOSPC49121-500MG APOSPC49121-250MG
(2,3-Difluorophenyl)methanesulphonyl chloride
(2,3-Difluorophenyl)methanesulphonyl chloride
Formula: C₇H₅ClF₂O₂S
Peso molecolare: 226,628 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06798045
CAS Number: 886498-49-3
UN: 3265
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR