Stampa…

Methyl 3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate

Fornitore: Apollo Scientific
Attenzione

PC48668-1G PC48668-250MG
APOSPC48668-1GEA 261 EUR
APOSPC48668-1G APOSPC48668-250MG
Methyl 3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Methyl 3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formula: C₁₁H₈F₃NO₂S
Peso molecolare: 275,247 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD27846135
CAS Number: 1609256-10-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR