Stampa…

6-Bromo-3-chloro-2-fluorobenzylamine

Fornitore: Apollo Scientific
Pericolo

Synonyms: (6-Bromo-3-chloro-3-fluorophenyl)methylamine

PC48561-1G PC48561-250MG
APOSPC48561-1GEA 437 EUR
APOSPC48561-1G APOSPC48561-250MG
6-Bromo-3-chloro-2-fluorobenzylamine
6-Bromo-3-chloro-2-fluorobenzylamine
Formula: C₇H₆BrClFN
Peso molecolare: 238,485 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28096624
CAS Number: 1499926-64-5
UN: 3259
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR