(4-Chlorooctafluorobutyl)trifluorooxirane
Fornitore: Apollo Scientific
|
Attenzione
|
PC450565-1G
PC450565-25G
PC450565-5G
APOSPC450565-1GEA
40.8
EUR
APOSPC450565-1G
APOSPC450565-25G
APOSPC450565-5G
(4-Chlorooctafluorobutyl)trifluorooxirane
(4-Chlorooctafluorobutyl)trifluorooxirane
Formula:
C₆ClF₁₁O Peso molecolare: 332,499 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD16619622 CAS Number: 50838-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...