Stampa…

Benzyl 1,1,2,2-tetrafluoroethyl ether

Fornitore: Apollo Scientific

PC450279-100G PC450279-25G PC450279-500G
APOSPC450279-100GEA 285 EUR
APOSPC450279-100G APOSPC450279-25G APOSPC450279-500G
Benzyl 1,1,2,2-tetrafluoroethyl ether
Benzyl 1,1,2,2-tetrafluoroethyl ether
Formula: C₉H₈F₄O
Peso molecolare: 208,153 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16620487
CAS Number: 115611-57-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR