1H,2H-1-Chloroperfluorocyclobutane
Fornitore: Apollo Scientific
Formula:
C₄H₂ClF₅ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD23134584 CAS Number: 1403667-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...