Stampa…

2,6-Difluoro-3-iodobenzoic acid 95+%

Fornitore: Apollo Scientific

PC446159-1G PC446159-5G
APOSPC446159-1GEA 87.3 EUR
APOSPC446159-1G APOSPC446159-5G
2,6-Difluoro-3-iodobenzoic acid 95+%
2,6-Difluoro-3-iodobenzoic acid
Formula: C₇H₃F₂IO₂
Peso molecolare: 283,999 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11040446
CAS Number: 229178-74-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR