Stampa…

5-Chloro-4-(chloromethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole 95+%

Fornitore: Apollo Scientific

PC446103-1G PC446103-500MG
APOSPC446103-1GEA 412 EUR
APOSPC446103-1G APOSPC446103-500MG
5-Chloro-4-(chloromethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole 95+%
5-Chloro-4-(chloromethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Formula: C₆H₅Cl₂F₃N₂
Peso molecolare: 233,019 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11105013
CAS Number: 1152623-73-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR